missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ST8SIA4 Recombinant Protein

Brand:  Abnova™ H00007903-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192182

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH27866
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
44.22
8
Glutathione Sepharose 4 Fast Flow
25 ug
MRSIRKKWTICTISLLLIFYKTKEIARTEEHQETQLIGDGELSLSRSLVNSSDKIIRKAGSSIFQHNVEGWKINSSLVLEIRKNILRFLDAERDVSVVKSSFKPGDVIHYVLDRRRTLNISHDLHSLLPEVSPMKNRRFKTCAVVGNSGILLDSECGKEIDSHNFVIR
MGC34450/MGC61459/PST/PST1/SIAT8D/ST8SIA-IV
ST8SIA4
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
7903
ST8SIA4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ST8SIA4
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers