missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Mouse anti-Human, Clone: ZB55, Novus Biologicals™
Mouse Monoclonal Antibody
483.00€
Specifications
| Antigen | ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 |
|---|---|
| Clone | ZB55 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Golgi Apparatus Markers | |
| 10 mM PBS with 0.05% BSA | |
| 6480 | |
| IgM κ | |
| PEG purified |
| ZB55 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N | |
| CD32 peptides | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title