missing translation for 'onlineSavingsMsg'
Learn More

SSX2IP Monoclonal Antibody (OTI1B6), TrueMAB™, OriGene

Product Code. 17527873
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17527873 30 μL 30µL
17507873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17527873 Supplier Origene Technologies Supplier No. TA807385S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSX2IP Monoclonal antibody specifically detects SSX2IP in Human samples. It is validated for Western Blot

Specifications

Antigen SSX2IP
Applications Western Blot
Classification Monoclonal
Clone OTI1B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PON3
Gene Accession No. Q15166
Gene Alias ADIP; Afadin- and alpha-actinin-binding protein; afadin- and alpha-actinin-binding protein ADIP; Afadin- and alpha-actinin-binding protein-like protein; afadin DIL domain-interacting protein; AU014939; AU042321; FLJ10848; KIAA0923; LOW QUALITY PROTEIN: afadin- and alpha-actinin-binding protein; MGC75026; SSX family member 2 interacting protein; SSX2-interacting protein; Ssx2ip; synovial sarcoma, X breakpoint 2 interacting protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 30-319 of human PON3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5446
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.