missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-4 Antibody (MC-813-70), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1435H
This item is not returnable.
View return policy
Description
SSEA-4 Monoclonal antibody specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| SSEA-4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| MC-813-70 | |
| Flow Cytometry, Immunocytochemistry | |
| SSEA-4,stage-specific embryonic antigen 4 | |
| 2120Ep human embryonal carcinoma cell line | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction