missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRY Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene
Description
SRY Monoclonal antibody specifically detects SRY in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
| Antigen | SRY |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI3C8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ASGR2 |
| Gene Accession No. | P07307 |
| Gene Alias | essential protein for sex determination in human males; HMG box transcription factor Sry1; HMG box transcription factor Sry3BI; sex determining region of Chr Y; sex determining region on Y; sex determining region Y; sex determining region Y protein; sex-determining region on Y; sex-determining region Y protein; SRXX1; SRXY1; Sry; Sry1; Sry3BI; SRYGENE; TDF; TDY; Testis determining locus; testis determining-Y; testis-determining factor; testis-determining factor on Y |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?