missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRRM1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93715-0.02ml
This item is not returnable.
View return policy
Description
SRRM1 Polyclonal antibody specifically detects SRRM1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SRRM1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin | |
| MGC39488, POP101, Ser/Arg-related nuclear matrix protein, serine/arginine repetitive matrix 1, serine/arginine repetitive matrix protein 1, SRm160, SRM160160-KD, SR-related nuclear matrix protein of 160 kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human SRRM1 (NP_005830.2). MDAGFFRGTSAEQDNRFSNKQKKLLKQLKFAECLEKKVDMSKVNLEVIKPWITKRVTEILGFEDDVVIEFIFNQLEVKNPDSKMMQINLTGFLNGKNAREFMGELWPLLLSAQENIAGIPSAFLELKKEEIKQRQIEQEKLASMKKQDEDKDKRDKEEKE | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10250 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction