Learn More
SRP9 Antibody (2D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43039
This item is not returnable.
View return policy
Description
SRP9 Monoclonal antibody specifically detects SRP9 in Human samples. It is validated for ELISA,Western Blot
Specifications
| SRP9 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6726 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Western Blot | |
| 2D4 | |
| Western Blot 1:1000 - 1:4000, ELISA | |
| ALURBP, MGC110981, signal recognition particle 9 kDa protein, signal recognition particle 9kD, signal recognition particle 9kDa | |
| signal recognition particle 9kDa - (Uniprot# P49458) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |