Description
SRI Polyclonal antibody specifically detects SRI in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | SRI |
| Conjugate | PE |
| Product Type | Antibody |
| Product Line | Novus |