missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Mouse anti-Human, Clone: SREBP1/1578, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | SREBP1 |
|---|---|
| Clone | SREBP1/1578 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA |
| Applications | Immunohistochemistry (Paraffin) |
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human samples. It is validated for ELISA.Specifications
| SREBP1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 6720 | |
| IgG1 κ | |
| Protein A or G purified |
| SREBP1/1578 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| Recombinant full-length human SREBP1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title