missing translation for 'onlineSavingsMsg'
Learn More

SRD5A2 Antibody, Novus Biologicals™

Product Code. p-7199863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18211704 0.025 mL 25µL
18294541 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18211704 Supplier Novus Biologicals Supplier No. NBP1465100.025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SRD5A2 Polyclonal specifically detects SRD5A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRD5A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1: 4000, Immunohistochemistry 1: 2000, Immunohistochemistry-Paraffin 1: 2000
Formulation Whole antisera with No Preservative
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase
Gene Symbols SRD5A2
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal region of human SRD5A2 (Steroid 5-alpha-reductase 2) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6716
Test Specificity Specific for SRD5A2.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.