missing translation for 'onlineSavingsMsg'
Learn More

Src Antibody (S05-7H8), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00€ - 645.00€

Specifications

Antigen Src
Clone S05-7H8
Concentration 0.3 mg/mL
Dilution Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18647934
View Documents
Novus Biologicals
NBP3-20055-100ul
100 μg
645.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18648354
View Documents
Novus Biologicals
NBP3-20055-50ul
50 μg
421.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Src Monoclonal antibody specifically detects Src in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Src
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
6714
IgG
Affinity purified
S05-7H8
Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
A synthetic peptide of human Src (Uniprot # P12931)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.