missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SQRDL Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93141-0.1ml
This item is not returnable.
View return policy
Description
SQRDL Polyclonal antibody specifically detects SQRDL in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SQRDL | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| CGI-44, sulfide dehydrogenase like, sulfide quinone reductase-like (yeast), sulfide:quinone oxidoreductase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 211-450 of human SQRDL (NP_067022.1). LSEAYFRKTGKRSKANIIFNTSLGAIFGVKKYADALQEIIQERNLTVNYKKNLIEVRADKQEAVFENLDKPGETQVISYEMLHVTPPMSPPDVLKTSPVADAAGWVDVDKETLQHRRYPNVFGIGDCTNLPTSKTAAAVAAQSGILDRTISVIMKNQTPTKKYDGYTSCPLVTGYNRVILAEFDYKAEPLETFPFDQSKERLSMYLMKADLMPFLYWNMMLRGYWGGPAFLRKLFHLGMS | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 58472 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction