missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY1 Monoclonal Antibody (669534)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561628
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Sprouty-1 and -2 are membrane-anchored phosphoprotein inhibitors of growth factor signaling in endothelial cells. Four mammalian Sprouty homologues (Sprouty-1-4) have been identified. One study showed that overexpressed Sprouty-1 and -2 inhibits fibroblast growth factor and vascular endothelial growth factor-induced proliferation and differentiation by repressing pathways leading to MAP kinase activation. Sprouty proteins are also endogenous inhibitors of the Ras/MAP kinase pathway that play an important role in the remodeling of branching tissues, such as in development of the lung, kidney tubules, vascular system, and breast ducts. A recent study shows that Sprouty-1 and -2 may have tumor suppressing activity in the breast, and found that both proteins are consistently down-regulated in breast carcinomas.
Specifications
| SPRY1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O43609 | |
| SPRY1 | |
| E.coli-derived recombinant human SPRY1, Met1-Lys178. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 669534 | |
| Unconjugated | |
| SPRY1 | |
| hSPRY1; Protein sprouty homolog 1; sprouty 1; sprouty homolog 1 (Drosophila); sprouty homolog 1, antagonist of FGF signaling; sprouty homolog 1, antagonist of FGF signaling (Drosophila); sprouty RTK signaling antagonist 1; sprouty, Drosophila, homolog of, 1 (antagonist of FGF signaling); sprouty1; SPRY1; spry-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction