missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRR1A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93464-0.1ml
This item is not returnable.
View return policy
Description
SPRR1A Polyclonal antibody specifically detects SPRR1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SPRR1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| 19 kDa pancornulin, cornifin A, cornifin-A, small proline-rich protein 1A, Small proline-rich protein IA, SPR-IA, SPRK | |
| A synthetic peptide corresponding to a sequence within amino acids 1-89 of human SPRR1A (NP_005978.2). CPSTVTPAPAQQKTKQK | |
| 0.1 mL | |
| Neuroscience | |
| 6698 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction