missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRED2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148909
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SPRED2 is a member of the Sprouty /SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (Nonami et al., 2004 ).
Specifications
| SPRED2 | |
| Polyclonal | |
| Unconjugated | |
| Spred2 | |
| C79158; Spred2; Spred-2; sprouty protein with EVH-1 domain 2, related sequence; sprouty related EVH1 domain containing 2; sprouty related, EVH1 domain containing 2; sprouty-related, EVH1 domain containing 2; sprouty-related, EVH1 domain-containing protein 2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 200734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q7Z698 | |
| Spred2 | |
| E. coli-derived recombinant human SPRED2, Lys117-Ser297. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction