missing translation for 'onlineSavingsMsg'
Learn More

SPPL2a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

231.00€ - 307.65€

Specifications

Antigen SPPL2a
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18611458
View Documents
Novus Biologicals
NBP3-06160-100ul
100 μg
ONLINE EXCLUSIVE PRICE
326.00€
307.65€ / 100µL
Save 18.35€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18676606
View Documents
Novus Biologicals
NBP3-06160-50ul
50 μg
231.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SPPL2a Polyclonal antibody specifically detects SPPL2a in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

SPPL2a
Immunofluorescence
Unconjugated
Rabbit
Cell Biology
PBS, pH7.0
84888
IgG
Antigen and protein A Affinity-purified
Immunocytochemistry/ Immunofluorescence 1:100-1:500
Polyclonal
Purified
RUO
Human
EC 3.4.23, EC 3.4.23.-, IMP-3, IMP3intramembrane cleaving protease, Intramembrane protease 3, Presenilin-like protein 2, PSL2, signal peptide peptidase-like 2A, SPPL2a, SPP-like 2A
Produced in rabbits immunized with E. coli-derived Human SPPL2a fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.