missing translation for 'onlineSavingsMsg'
Learn More

SPOP Antibody (S01-5B7), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00€ - 645.00€

Specifications

Antigen SPOP
Clone S01-5B7
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676423
View Documents
Novus Biologicals
NBP3-19781-100ul
100 μg
645.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18647663
View Documents
Novus Biologicals
NBP3-19781-50ul
50 μg
421.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SPOP Monoclonal antibody specifically detects SPOP in Human samples. It is validated for Western Blot
Specifications

Specifications

SPOP
0.3 mg/mL
Monoclonal
Purified
RUO
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
8405
IgG
Affinity purified
S01-5B7
Western Blot
Unconjugated
Rabbit
Human
HIB homolog 1, Roadkill homolog 1, speckle-type POZ protein, TEF2
A synthetic peptide of human SPOP (Uniprot # O43791)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.