missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK5 Antibody (S06-6C1), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Specifications
| Antigen | SPINK5 |
|---|---|
| Clone | S06-6C1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628493
|
Novus Biologicals
NBP3-19779-50ul |
50 μg |
421.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689093
|
Novus Biologicals
NBP3-19779-100ul |
100 μg |
645.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPINK5 Monoclonal antibody specifically detects SPINK5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SPINK5 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Kazal type 5, serine peptidase inhibitor, Kazal type 5 | |
| A synthetic peptide of human SPINK5 (Uniprot # Q9NQ38) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-6C1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cardiovascular Biology, Cell Biology | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 11005 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.