missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sphingosine Kinase 2/SPHK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30877-100ug
This item is not returnable.
View return policy
Description
Sphingosine Kinase 2/SPHK2 Polyclonal antibody specifically detects Sphingosine Kinase 2/SPHK2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Sphingosine Kinase 2/SPHK2 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL, ELISA | |
| EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2 | |
| Synthetic peptide derived from the N-terminal extension of the Sphingosine Kinase 2/SPHK2 protein. | |
| 100 μg | |
| Apoptosis | |
| 56848 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction