missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93512-0.02ml
This item is not returnable.
View return policy
Description
SPG3A Polyclonal antibody specifically detects SPG3A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SPG3A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| AD-FSP, atlastin GTPase 1, atlastin1, atlastin-1, Brain-specific GTP-binding protein, EC 3.6.5.-, FSP1, GBP3, GBP-3, GTP-binding protein 3, Guanine nucleotide-binding protein 3, guanylate-binding protein 3, Spastic paraplegia 3 protein A, spastic paraplegia 3A (autosomal dominant), SPG3, SPG3AhGBP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ATL1 (NP_056999.2). MAKNRRDRNSWGGFSEKTYEWSSEEEEPVKKAGPVQVLIVKDDHSFELDETALNRILLSEAVRDKEVVAVSVAGAFRKGKSFLMDFMLRYMYNQESVDWVGDYNEPLTGFSWRGGSERETTGIQIWSEIFLINKPDGKKVAVLLMDTQGTFDSQSTLRDSATVFALSTMISSIQVYNLSQNVQEDDLQHLQLFTEYGRLAMEETFLKPFQSLIFLVRDWSFPYEFSYGADGGAKFLEKRLKVSGNQHEELQNVRKHIHSCFTNISCFLLPHPGLKVATNP | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51062 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction