Learn More
Cole-Parmer™ Replacement Cylinders for Mid-Size Grinding Vial Sets
Product Code. 15330482 Shop All SPEX Certiprep Products
Change view
Click to view available options
For Use With (Equipment):
Mid Size, Cr-Free, Poly-Vial Grinding Vial Set
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15330482 Mid Size, Cr-Free, Poly-Vial Grinding Vial Set Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15330482 Supplier Cole-Parmer™ Supplier No. 6881C4
This item requires a price update prior to being ordered.
Request a Price for this item here.
Replacement Cylinders for 6881 mid-size grinding vial set, 6883 mid-size low-chrome grinding vial set, and 6885 Mid-size poly-vial set.
  • Withstands liquid nitrogen temperatures.
  • Are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Transparent plastic center cylinder allows visual checking of the grinding progress.
Specifications
Product Type Center Cylinder
For Use With (Equipment) Mid Size, Cr-Free, Poly-Vial Grinding Vial Set
Material Polycarbonate
Packaging Quantity 4 Pk.