Learn More
Cole-Parmer™ Grinding Vial Set, Mid-Size Vial, PC, Low-Chrome Steel
Product Code. 15370482 Shop All SPEX Certiprep Products
Click to view available options
Material:
Polycarbonate
Unit Size:
1 set
Product Code. 15370482
Brand: Cole-Parmer™ 6883
This item requires a price update prior to being ordered.
Request a Price for this item here.
Requires
886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
Mid-Size Grinding Vial for Freezer/Mill™ cryogenic grinders to simplify your processes.
  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Vial sets consist of a center cylinder, two end plugs, and an impactor.
  • Specifically designed to grind electronic components for RoHS/WEEE compliance testing.
  • Withstands liquid nitrogen temperatures.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Specifications
Product Type Grinding Set
Capacity (English) 0.10 to 0.88 oz.
Capacity (Metric) 3 to 25 g
For Use With (Equipment) Large Freezer/Mill (Mfr. No. 6875) and Large Dual Freezer/Mill (Mfr. No. 6875 D), When Used with Mid-Size Grinding Vial Adapter (6887)
Includes 1 6883 P Steel Impactor, 2 6883 E Steel End Plugs, 1 Pk. Center Cylinders (6811C4)
Material Polycarbonate
Material (Secondary) Low-chrome Steel
Size Mid-size