Learn More
Cole-Parmer™ Replacement End Plugs for Large Grinding Vial Set, Stainless Steel
Product Code. 12588706 Shop All SPEX Certiprep Products
Change view
Click to view available options
Packaging Quantity:
2 Pk.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
12588706 2 Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12588706 Supplier Cole-Parmer™ Supplier No. 6801E
This item requires a price update prior to being ordered.
Request a Price for this item here.
Replacement end plugs for 6801 Large Grinding Vial Set.
  • Made of 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Product Type End Plug
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) 6801 Large Grinding Vial Set
Material Stainless Steel (440 C)
Packaging Quantity 2 Pk.