Learn More
SPEX™ SamplePrep Grinding Vial Accessory Packages
Product Code. 13476148 Shop All SPEX Certiprep Products
Change view
Click to view available options
Size:
Small
Large
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13476148 Large Each
15330502 Small 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13476148 Supplier SPEX™ SamplePrep Supplier No. 6870L
This item requires a price update prior to being ordered.
Request a Price for this item here.
Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™
  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Compatible with grinding vial sets 61043-09 and 61043-11.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Specifications
Product Type Grinding Set
For Use With (Equipment) Freezer/Mill Cryogenic Grinders
Includes 1 Vial Extractor/Opener (04576-38) and Vial Rack (04576-39)
Size Large