Learn More
Cole-Parmer™ Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer
Product Code. 12587326 Shop All SPEX Certiprep Products
Change view
Click to view available options
Holds:
15 x 15 mL Polycarbonate Cryovials
24 x 5 mL Polycarbonate Vials
24 x 5 mL Polyethylene Vials
48 x 1.5 to 2 mL Microcentrifuge or PCR Tubes
6 x 15 mL Short Polycarbonate Vials
6 x 50 mL PC or PE Cryovials
Unit Size:
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
12587326 15 x 15 mL Polycarbonate Cryovials Pack of 2
15360462 24 x 5 mL Polycarbonate Vials Pack of 2
15350462 6 x 15 mL Short Polycarbonate Vials Pack of 2
15390462 24 x 5 mL Polyethylene Vials Pack of 2
15370462 48 x 1.5 to 2 mL Microcentrifuge or PCR Tubes Pack of 2
12597326 6 x 50 mL PC or PE Cryovials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12587326 Supplier Cole-Parmer™ Supplier No. 2661
This item requires a price update prior to being ordered.
Request a Price for this item here.
These Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Product Type Holder
For Use With (Application) Ideal for High Throughput Cryogenic Grinding and to Preserve Pesticides, RNA, Proteins and Other Temperature Sensitive Samples
For Use With (Equipment) Spex Geno/Grinder
Holds 15 x 15 mL Polycarbonate Cryovials
Material Aluminum