Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Product Code. 15390532 Shop All SPEX Certiprep Products
Change view
Click to view available options
Product Type:
Clamps for slurry grinding
Multiple Sample Adapter
Vial Clamp Adapter
Holds:
4 x 0.75 in. Vials
7 x 0.5 in. dia. Vials
Alumina Ceramic Grinding Vial Set (41005-86)
Vial Adapters (04576-65 and 04576-66)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Holds unitSize
15390532 Multiple Sample Adapter 4 x 0.75 in. Vials Each
12564236 Multiple Sample Adapter 7 x 0.5 in. dia. Vials Each
15340522 Clamps for slurry grinding Alumina Ceramic Grinding Vial Set (41005-86) Each
15300542 Vial Clamp Adapter Vial Adapters (04576-65 and 04576-66) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15390532 Supplier Cole-Parmer™ Supplier No. 8011SSP
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use adapters to run multiple samples or to hold vial securely in place during grinding.
  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Product Type Multiple Sample Adapter
For Use With (Application) Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples
For Use With (Equipment) 8000 D and 8000 M Mixer or Mill
Holds 4 x 0.75 in. Vials