Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Shop All SPEX Certiprep ProductsDescription
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Specifications
| Product Type | Vial Clamp Adapter |
| For Use With (Application) | Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples |
| For Use With (Equipment) | 8000 D and 8000 M Mixer or Mill |
| Holds | Vial Adapters (04576-65 and 04576-66) |
| Material | Plastic |