Learn More
Cole-Parmer™ Vial Set for High-Energy Mixer/Mill™ Grinders
Product Code. 15349681 Shop All SPEX Certiprep Products
Click to view available options
Material:
Agate
Alumina Ceramic
Hardened Steel
Stainless Steel
Tungsten Carbide-lined
Zirconia
Diameter (Metric):
1.27 cm
1.9 cm
5.7 cm
5.71 cm
5.72 cm
6.03 cm
6.35 cm
Height (Metric):
2.54 cm
4.76 cm
5.08 cm
6.83 cm
6.98 cm
7.62 cm
Unit Size:
1 set
Each
Product Code. 15349681
Brand: Cole-Parmer™ 8001
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use this Vial Set with high-energy Mixer/Mill™ grinders to pulverize and mix samples.
  • Crush, grind, and mill various materials.
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinders with a wide range of vial sets suitable for your application.
Specifications
Product Type Grinding Vial Set
Diameter (English) 2.25 in.
Diameter (Metric) 5.72 cm
For Use With (Equipment) 8000 D, 8000 M Mixer/Mills
Height (English) 3 in.
Height (Metric) 7.62 cm
Includes Two 1/2 in. and Four 1/4 in. Steel Grinding Balls
Material Hardened Steel