Learn More
SPEX CertiPrep™ OdorEroder
Product Code. p-4186263 Shop All SPEX Certiprep Products
Click to view available options
Volume (Metric):
100 g
250 g
Unit Size:
100g
250g
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15286106
Brand: SPEX CertiPrep™ ODER100G
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Effectively neutralizes a wide range of chemical odors in the lab
  • Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
  • As odors get trapped, the beads start turning brown
Specifications
For Use With (Application) Effectively neutralizes a wide range of chemical odors in the lab
Volume (Metric) 100 g