missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrometric Oil Standard D12-030: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 30 μg/g in Aviation Reference Oil, VHG™
Click to view available options
Quantity:
240 mL
Unit Size:
240mL
Description
Spectrometric Oil Standard D12-030: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 30 μg/g in Aviation Reference Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| CAS | 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4 |
| Methods | RDE |
| Analyte or Component Names | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Aviation Oil |
| Quantity | 240 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 30 μg/g/Chromium 30 μg/g/Copper 30 μg/g/Iron 30 μg/g/Lead 30 μg/g/Magnesium 30 μg/g/Nickel 30 μg/g/Silicon 30 μg/g/Silver 30 μg/g/Sodium 30 μg/g/Tin 30 μg/g/Titanium 30 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction