Learn More
SPE tube phenyl reversed phase polypropylene 500mg, 3mL Bakerbond
Product Code. 10492991 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10492991 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10492991 Supplier JT Baker Supplier No. 709503
  • ONLINE EXCLUSIVE PRICE
    482.45€
    390.49€ / Pack of 50
    Save 91.96€
    19% Off
 Restricted Item
Estimated Shipment: 04-03-2027