Learn More
SPE tube ODS reversed phase glass 2g, 8mL Bakerbond
Brand: JT Baker 7334-08
646.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : CAS : 71889-02-6 Weight : 0.60000kg
Brand: JT Baker 7334-08
646.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : CAS : 71889-02-6 Weight : 0.60000kg