Learn More
SPE tube Florisil normal phase polypropylene 500mg, 3mL Bakerbond
Brand:  JT Baker 7213-03
247.95 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : CAS : 1343-88-0 Weight : 0.10000kg
Product Code. 10782422
  • 261.00€ / Pack of 50
Estimated Shipment: 23-07-2024