missing translation for 'onlineSavingsMsg'
Learn More

SPC25 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16844935 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16844935 20 μL 20µL
16834935 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16844935 Supplier Proteintech Supplier No. 264741AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein that may be involved in kinetochore-microtubule interaction and spindle checkpoint activity.

Specifications

Antigen SPC25
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SPC25
Gene Accession No. Q9HBM1
Gene Alias AD024, hSpc25, Kinetochore protein Spc25, SPBC25, SPC25
Gene Symbols SPC25
Host Species Rabbit
Immunogen SPC25 Fusion Protein Ag23985
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57405
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.