missing translation for 'onlineSavingsMsg'
Learn More

SPATA22 Antibody (OTI6H9) - Azide and BSA Free, Novus Biologicals™

Product Code. 18197256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18197256 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18197256 Supplier Novus Biologicals Supplier No. NBP274309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPATA22 Monoclonal specifically detects SPATA22 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA22
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6H9
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20
Gene Symbols SPATA22
Host Species Mouse
Immunogen Full length human recombinant protein of human SPATA22(NP_115987) produced in E.coli.
Molecular Weight of Antigen 41.1 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 84690
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.