missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spastin Mouse anti-Human, Mouse, Rat, Clone: Sp 6C6, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | Spastin |
|---|---|
| Clone | Sp 6C6 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Western Blot, Immunofluorescence |
Description
Spastin Monoclonal specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Spastin | |
| 0.2 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4 | |
| Recombinant full-length human Spastin protein. | |
| Primary | |
| Store at 4°C. |
| Sp 6C6 | |
| Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6683 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title