missing translation for 'onlineSavingsMsg'
Learn More

Spartin, SPG20 Rabbit anti-Human, Mouse, Rat, CoraLite 647, Polyclonal, Proteintech

Product Code. 17771459 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17771459 20 μL 20µL
17649986 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17771459 Supplier Proteintech Supplier No. CL6471379120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Spartin, SPG20 Polyclonal antibody apecifically detetects Spartin, SPG20 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.

Specifications

Antigen Spartin, SPG20
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 ug/mL
Conjugate CoraLite 647
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC047083
Gene Symbols SPG20
Host Species Rabbit
Immunogen Spartin, SPG20 fusion protein Ag4815
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 23111
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.