Learn More
Duran™ Spare Glass Pipettes
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Pack of 10
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Color | Clear |
| Material | Glass |
| Quantity | 10Pack |
| Product Type | Spare Pipette |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur