missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP110 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76385-0.025mg
This item is not returnable.
View return policy
Description
SP110 Polyclonal specifically detects SP110 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SP110 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FLJ22835, IFI41, IFI75, interferon-induced protein 41, 30kD, Interferon-induced protein 41/75, interferon-induced protein 75, 52kD, IPR1, phosphoprotein 41, phosphoprotein 75, SP110 nuclear body protein, Speckled 110 kDa, Transcriptional coactivator Sp110, VODI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3431 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_004501 | |
| SP110 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of the human IPR1. The immunogen is located within amino acids 70 - 120 of IPR1. Amino Acid Squence: QINLREYPNLVTIYRS | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction