missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP-D Antibody (S06-2B1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19975-100ul
This item is not returnable.
View return policy
Description
SP-D Monoclonal antibody specifically detects SP-D in Mouse, Rat samples. It is validated for Western Blot
Specifications
| SP-D | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-2B1 | |
| Unconjugated | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| Recombinant protein of mouse SP-D (Uniprot # P50404) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 6441 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction