missing translation for 'onlineSavingsMsg'
Learn More

SOX2 Mouse anti-Human, Mouse, Clone: rSOX2/1792, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07888-100ug

 View more versions of this product

Product Code. 18694189

  • 499.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOX2 Monoclonal specifically detects SOX2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOX2
Monoclonal
0.2 mg/mL
ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary)
100 μg
Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
6657
Store at 4°C.
IgG1 κ
Western Blot, Immunohistochemistry (Paraffin)
rSOX2/1792
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.