missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX11 Antibody (CL0143) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52865CL1
This item is not returnable.
View return policy
Description
SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SOX11 | |
| Monoclonal | |
| CoraFluor 1 | |
| SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 | |
| This antibody was developed using a recombinant protein derived from P35716, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0143 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6664 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction