missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Mouse anti-Human, Mouse, Clone: SOX10/1074, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14443.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human SOX10.
- High specificity
- High reactivity
Specifications
| SOX10 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end u | |
| P56693 | |
| SOX10 | |
| Recombinant protein corresponding to amino acids 115-269 of human SOX10. | |
| 100 μg | |
| Primary | |
| Recognizes a protein of ∼55kDa, identified as SOX10. This antibody is highly specific and does not cross-react with other members of the SOX-family. | |
| Store at 4°C. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| SOX10/1074 | |
| Mouse monoclonal antibody raised against partial recombinant human SOX10. | |
| In 10 mM PBS (0.05% BSA and 0.05% azide). | |
| DOM/MGC15649/WS2E/WS4 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 6663 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction