missing translation for 'onlineSavingsMsg'
Learn More

SOX10 Antibody (CL4455) [FITC], Novus Biologicals™

Product Code. 30305489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30305489 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30305489 Supplier Novus Biologicals Supplier No. NBP259050F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX10 Monoclonal antibody specifically detects SOX10 in Feline,Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX10
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL4455
Conjugate FITC
Formulation PBS
Gene Alias MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P56693, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6663
Target Species Feline, Human, Mouse
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.