missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody (CL4455) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59050G
This item is not returnable.
View return policy
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Feline,Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| SOX10 | |
| Monoclonal | |
| DyLight 488 | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| This antibody was developed using a recombinant protein derived from P56693, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Feline, Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| CL4455 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6663 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction