Description
SOX10 Monoclonal antibody specifically detects SOX10 in Feline,Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | SOX10 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL4455 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P56693, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |