missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sortilin Antibody (CL6528) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76501G
This item is not returnable.
View return policy
Description
Sortilin Monoclonal antibody specifically detects Sortilin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Sortilin | |
| Monoclonal | |
| DyLight 488 | |
| 100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1 | |
| This antibody was developed using a recombinant protein derived from Q99523, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 6272 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL6528 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction