Learn More
Sortilin Antibody (CL6528) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76501APC
Product Code. 30290569
  • 737.00€ / 0.10mL
Description
Sortilin Monoclonal antibody specifically detects Sortilin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sortilin
Monoclonal
APC
100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1
This antibody was developed using a recombinant protein derived from Q99523, with the exact immunogen sequence remaining proprietary.
0.1 mL
Golgi Apparatus Markers
6272
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL6528
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse