missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R5/SSTR5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-74540-0.025ml
This item is not returnable.
View return policy
Description
Somatostatin R5/SSTR5 Polyclonal specifically detects Somatostatin R5/SSTR5 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| Somatostatin R5/SSTR5 | |
| Polyclonal | |
| Western Blot 1:4000 - 1:6000, Flow Cytometry 3 - 5 μg 106 cells, Immunohistochemistry-Paraffin 1:2500, Immunocytochemistry 1:5000 | |
| P35346 | |
| SSTR5 | |
| Synthetic peptide corresponding to residues Q(344) E A T R P A H R A A A N G L M Q T S K L(364) of human SSTR5. | |
| 0.025 mL | |
| Primary | |
| Somatostatin Receptor 5 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 6755 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction