missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R3/SSTR3 Antibody (7H8E5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
187.00€ - 562.00€
Specifications
| Antigen | Somatostatin R3/SSTR3 |
|---|---|
| Clone | 7H8E5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602776
|
Novus Biologicals
NBP2-37266-0.025ml |
0.025 mL |
187.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18113639
|
Novus Biologicals
NBP2-37266 |
0.1 mL |
562.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin R3/SSTR3 Monoclonal specifically detects Somatostatin R3/SSTR3 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Somatostatin R3/SSTR3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| somatostatin receptor 3, SS-3-R, SS3-R | |
| SSTR3 | |
| IgG1 | |
| Affinity Purified |
| 7H8E5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Breast Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS with 0.05% Sodium Azide | |
| 6753 | |
| Purified recombinant fragment of human SSTR3 (AA: 1-43) expressed in E. Coli. | |
| Primary | |
| 45.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title